Få mere at vide
B Braun™ Sterican™ Insulin Syringe Needles
Beskrivelse
- Needle made of stainless chromium-nickel steel with extremely smooth surface and light silicone coating for minimal pain upon puncture.
- Transparent Luer Lock plastic hub made of polypropylene
- Colour-coded hub identifies the outer diameter of the needle, conforms to ISO 6009
- Latex-free/PVC-free
Tekniske data
Tekniske data
| Autoklaverbar | Autoclavable |
| Certificeringer/compliance | ISO 6009 |
| Nålemåler | 30 G |
| Diameter (metrisk) Udvendig | 0.30 mm |
| Længde (metrisk) | 12 mm |
| Mængde | 100/Pk. |
| Emballage | Sterile, single-packed |
| Sterilitet | Sterile |