Få mere at vide
Inosine-5'-monophosphate Sodium Salt Hydrate, TRC
Artikelnummer. 30010991
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010991 500 mg
30008004 100 mg
30006355 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010991 Leverandør Toronto Research Chemicals Leverandørnr. TRCI665010500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 20813-76-7
Molekylær formel C10 H11 N4 O8 P . 2[Na] . 8[H2 O]
Molekylvægt (g/mol) 536.292
IUPAC navn disodium;[(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-1H-purin-9-yl)oxolan-2-yl]methyl phosphate;octahydrate
SMIL O.O.O.O.O.O.O.O.[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)([O-])[O-])n2cnc3C(=O)NC=Nc23
Tekniske data
Kemisk navn eller materiale Inosine-5'-monophosphate Sodium Salt Hydrate
Mængde 500 mg
InChI formel InChI=1S/C10H13N4O8P.2Na.8H2O/c15-6-4(1-21-23(18,19)20)22-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;;;;;;;;;;/h2-4,6-7,10,15-16H,1H2,(H,11,12,17)(H2,18,19,20);;;8*1H2/q;2*+1;;;;;;;;/p-2/t4-,6-,7-,10-;;;;;;;;;;/m1........../s1
Formel vægt 536.096