missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Inosine-5'-monophosphate Disodium Salt Hydrate, TRC
Specifications
Specifications
| Kemisk navn eller materiale | 5'-Inosinic acid disodium salt hydrate |
| CAS | 352195-40-5 |
| Molekylær formel | C10 H11 N4 O8 P . 2[Na] . x[H2 O] |
| Mængde | 5 g |
| Synonym | 5'-Inosinic acid, disodium salt, hydrate (9CI), 5'-Inosinic Acid Disodium Salt Hydrate |
| InChI formel | InChI=1S/C10H13N4O8P.2Na.H2O/c15-6-4(1-21-23(18,19)20)22-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;;;/h2-4,6-7,10,15-16H,1H2,(H,11,12,17)(H2,18,19,20);;;1H2/q;2*+1;/p-2/t4-,6-,7-,10-;;;/m1.../s1 |
| SMIL | O.[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)([O-])[O-])n2cnc3C(=O)NC=Nc23 |
| IUPAC navn | disodium;[(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-1H-purin-9-yl)tetrahydrofuran-2-yl]methyl phosphate;hydrate |
| Molekylvægt (g/mol) | 410.18 |
| Formel vægt | 410.0216 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?