Få mere at vide
Ingliforib, MedChemExpress
Artikelnummer. 16497758
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16497758 10 mg 10mg
16483727 50 mg -
16447616 100 mg -
16430228 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16497758 Leverandør Medchem Express Leverandørnr. HY19396
MedChemExpress Ingliforib (CP 368296) is a glycogen phosphorylase inhibitor, with IC50s of 52, 352 and 150 nM for liver, muscle and brain glycogen phosphorylase, and has cardioprotective activity.
Ingliforib (CP 368296) is a glycogen phosphorylase inhibitor, with IC50s of 52, 352 and 150 nM for liver, muscle and brain glycogen phosphorylase, and has cardioprotective activity.
Tekniske data
Kemisk navn eller materiale Ingliforib
CAS 186392-65-4
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C23H24ClN3O5
Mængde 10 mg
Synonym CP 368296 GPi 296
Opløselighedsinformation DMSO : 130 mg/mL (283.90 mM; Need ultrasonic)
SMIL O=C(C(N1)=CC2=C1C=CC(Cl)=C2)N[C@@H](CC3=CC=CC=C3)[C@@H](O)C(N4C[C@@H](O)[C@@H](O)C4)=O
Molekylvægt (g/mol) 457.91
Formel vægt 457.91
Procent renhed 99.64%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation