missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
ING5 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06217C
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
ING5 Polyclonal specifically detects ING5 in Human samples. It is validated for Western Blot, ELISA.
Tekniske data
| ING5 | |
| Polyclonal | |
| Western Blot, ELISA | |
| inhibitor of growth family, member 5, inhibitor of growth protein 5, p28ING5FLJ23842 | |
| Produced in rabbits immunized with purified, recombinant Human ING5 (Q8WYH8-2; Glu36-Gln84). | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 84289 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion