missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Influenza A Hemagglutinin H1 Antibody (InA139), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-73160F
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Influenza A Hemagglutinin H1 Monoclonal antibody specifically detects Influenza A Hemagglutinin H1 in Virus samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Tekniske data
| Influenza A Hemagglutinin H1 | |
| Monoclonal | |
| FITC | |
| HA, Hemagglutinin, Hemagglutinin HA | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/NewCaledonia/20/99 (strain H1N1). | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Sandwich ELISA | |
| InA139 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion