missing translation for 'onlineSavingsMsg'
Få mere at vide

Indomethacin Acyl-beta-D-glucuronide (>90%), TRC

Product Code. 30134352
Change view
Click to view available options
Mængde:
1 mg
2.5 mg
5 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
30134352 5 mg -
30138571 1 mg -
30012444 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134352 Supplier Toronto Research Chemicals Supplier No. TRCI6410105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75523-11-4
Molekylær formel C25 H24 Cl N O10
Molekylvægt (g/mol) 533.91
Synonym Indomethacin Acyl-B-D-Glucuronide
IUPAC navn (2S,3S,4S,5R,6S)-6-[2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMIL COc1ccc2c(c1)c(CC(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)c(C)n2C(=O)c4ccc(Cl)cc4

Specifications

Kemisk navn eller materiale Indomethacin Acyl-beta-D-glucuronide
Mængde 5 mg
InChI formel InChI=1S/C25H24ClNO10/c1-11-15(10-18(28)36-25-21(31)19(29)20(30)22(37-25)24(33)34)16-9-14(35-2)7-8-17(16)27(11)23(32)12-3-5-13(26)6-4-12/h3-9,19-22,25,29-31H,10H2,1-2H3,(H,33,34)/t19-,20-,21+,22-,25+/m0/s1
Formel vægt 533.1089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.