missing translation for 'onlineSavingsMsg'
Learn More

Indole-4-boronic acid pinacol ester, 97+%, Thermo Scientific™

Catalog Number 15438977
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
388116-27-6
C14H18BNO2
243.11
MFCD08689896
QDCIXBBEUHMLDN-UHFFFAOYSA-N
indole-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1h-indole-4-boronic acid, pinacol ester, 1h-indole, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, indole-4-boronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3-dioxaborolan-2-yl-1h-indole, 4-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl-1h-indole, pubchem19977
11322471
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
CC1(C)OB(OC1(C)C)C1=C2C=CNC2=CC=C1
This item is not returnable. View return policy
388116-27-6
C14H18BNO2
243.11
MFCD08689896
QDCIXBBEUHMLDN-UHFFFAOYSA-N
indole-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1h-indole-4-boronic acid, pinacol ester, 1h-indole, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, indole-4-boronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3-dioxaborolan-2-yl-1h-indole, 4-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl-1h-indole, pubchem19977
11322471
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
CC1(C)OB(OC1(C)C)C1=C2C=CNC2=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 243.11
Percent Purity ≥97%
Chemical Name or Material Indole-4-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'