Få mere at vide
Indole-3-acetic-alpha,alpha-d2 Acid, CDN
Artikelnummer. 30328508
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328508 0.05 g
30321610 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328508 Leverandør CDN Leverandørnr. CDND17090.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1 H-Indole-3-acetic acid carboxymethyl-d2
CAS 24420-86-8
Molekylær formel C10 2H2 H7 N O2
Mængde 0.05 g
Synonym 1 H-Indole-3-acetic acid D2 (carboxymethyl-D2), 1 H-Indole-3-acetic-α,α-d2 acid, Indole-3-acetic-α,α-d2 acid (8 CI), 2,2-Dideuterio-2-(1 H-indol-3-yl)acetic acid, 2-(3-Indolyl)acetic-2,2-d2 acid, 3-Indolylacetic acid-d2
InChI formel InChI=1 S/C10H9NO2/c12-10(13)5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6,11 H,5H2,(H,12,13)/i5D2
SMIL [2 H]C([2 H])(C(=O)O)c1c[nH]c2ccccc12
IUPAC navn 2,2-dideuterio-2-(1 H-indol-3-yl)acetic acid
Molekylvægt (g/mol) 177.2
Formel vægt 177.0759 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity