Få mere at vide
Indole-2,4,5,6,7-d5, TRC
Artikelnummer. 30403891
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30403891 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403891 Leverandør Toronto Research Chemicals Leverandørnr. TRCI577324100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Indole-2,4,5,6,7-d5
CAS 73509-21-4
Molekylær formel C8H2D5N
Mængde 100 mg
Synonym 1H-Indole-2,4,5,6,7-d5 (9CI), Indole-2,4,5,6,7-d5, Indole-d5, Indole-2,4,5,6,7-D5
InChI formel InChI=1S/C8H7N/c1-2-4-8-7(3-1)5-6-9-8/h1-6,9H/i1D,2D,3D,4D,6D
SMIL [2H]c1cc2c([2H])c([2H])c([2H])c([2H])c2[nH]1
IUPAC navn 2,4,5,6,7-pentadeuterio-1H-indole
Molekylvægt (g/mol) 122.18
Formel vægt 122.0892