Få mere at vide
Indole-2,4,5,6,7-d5-3-acetamide, CDN
Artikelnummer. 30323432
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323432 0.05 g
30321491 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323432 Leverandør CDN Leverandørnr. CDND66060.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1 H-Indole-d5-3-acetamide
CAS 1204700-53-7
Molekylær formel C10 2H5 H5 N2 O
Mængde 0.05 g
InChI formel InChI=1 S/C10H10N2O/c11-10(13)5-7-6-12-9-4-2-1-3-8(7)9/h1-4,6,12 H,5H2,(H2,11,13)/i1D,2 D,3 D,4 D,6 D
SMIL [2 H]c1[nH]c2c([2 H])c([2 H])c([2 H])c([2 H])c2c1CC(=O)N
IUPAC navn 2-(2,4,5,6,7-pentadeuterio-1 H-indol-3-yl)acetamide
Molekylvægt (g/mol) 179.23
Formel vægt 179.1107 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity