missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indoine Blue, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Infrared Spectrum | Authentic |
IUPAC Name | 3-amino-2,8-dimethyl-7-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride |
Lambda Max | 595 to 601nm (0.001% in methanol) |
Formula Weight | 506.01 |
Loss on Drying | 10% max. (110°C, 1 h) |
Quantity | 1 g |
Chemical Name or Material | Indoine Blue |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'