Få mere at vide
Indigo Carmine 5,7'-Isomer (~90%), TRC
Artikelnummer. 17963875
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17963875 50 mg
17914865 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17963875 Leverandør Toronto Research Chemicals Leverandørnr. TRCI52117550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27414-68-2
Molekylær formel C16 H8 N2 Na2 O8 S2
Molekylvægt (g/mol) 466.35
Synonym 2-(1,3-Dihydro-3-oxo-7-sulfo-2H-indol-2-ylidene)-2,3-dihydro-3-oxo-1H-indole-5-sulfonic Acid Sodium Salt, 3,3'-Dioxo-[δ2,2'-Biindoline]-5,7'-disulfonic Acid Disodium Salt
IUPAC navn [(2E)-3-oxo-2-(3-oxo-5-sodiooxysulfonyl-indolin-2-ylidene)indolin-7-yl]sulfonyloxysodium
SMIL [Na]OS(=O)(=O)c1ccc2N\C(=C/3\Nc4c(cccc4S(=O)(=O)O[Na])C3=O)\C(=O)c2c1
Tekniske data
Kemisk navn eller materiale Indigo Carmine 5,7'-Isomer
Mængde 50 mg
InChI formel InChI=1S/C16H10N2O8S2.2Na/c19-15-8-2-1-3-11(28(24,25)26)12(8)18-14(15)13-16(20)9-6-7(27(21,22)23)4-5-10(9)17-13;;/h1-6,17-18H,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2/b14-13+;;
Formel vægt 465.9517