Learn More
QIAGEN QIAcard FTA Indicating Classic
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Classic with 4 sample areas per card.
Brand:  QIAGEN WB120206
Additional Details : Weight : 0.61200kg
 View more versions of this product
Product Code. 10233352
  • 4645.00 DKK / Pack of 100
Estimated Shipment: 15-05-2024
to see stock.
Description
Description
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Classic with 4 sample areas per card.
  • QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
  • Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
  • Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Specifications
Collect, stabilize, process, transport, and archive buccal DNA samples, four samples per card.
Buccal DNA
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
4
100 Pk.
Card
Direct STR amplification
Pink indicating dye
Indicating FTA Classic Card
125 μL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning