Få mere at vide
Indeno[1,2,3-c,d]pyrene-d12, CDN
Artikelnummer. 30323614
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323614 0.01 g
30319465 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323614 Leverandør CDN Leverandørnr. CDND42490.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Indeno(1,2,3-c,d)pyrene D12
CAS 203578-33-0
Molekylær formel C22 2H12
Mængde 0.01 g
Synonym Indeno[1,2,3-cd]pyrene-1,2,3,4,5,6,7,8,9,10,11,12-d12, Indeno[1,2,3-cd]pyrene-d12 (9 CI), Indeno[1,2,3-c,d]pyrene D12, Indeno(1,2,3-c,d)pyrene D12
InChI formel InChI=1 S/C22H12/c1-2-7-17-16(6-1)18-11-10-14-9-8-13-4-3-5-15-12-19(17)22(18)21(14)20(13)15/h1-12 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D
SMIL [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c3c([2 H])c([2 H])c4c([2 H])c([2 H])c5c([2 H])c([2 H])c([2 H])c6c([2 H])c2c3c4c56
IUPAC navn Indeno[1,2,3-cd]pyrene-1,2,3,4,5,6,7,8,9,10,11,12-d12
Molekylvægt (g/mol) 288.4
Formel vægt 288.1692 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity