missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
INCENP Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29419-1-AP-150UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The INCENP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human INCENP. This antibody recognizes human antigen. The INCENP antibody has been validated for the following applications: WB, ELISA analysis.Tekniske data
| INCENP | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3619 | |
| Store at -20°C. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, ELISA | |
| 800 μg/mL | |
| FLJ31633; MGC111393 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human | |
| 106-130 kDa | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion