Få mere at vide
DiscoverX InCELL Hunter™ HEK 293 BLK Kinase Binding Cell Line
Artikelnummer. 17394779
Encompass
Mængde:
2 hætteglas
Artikelnummer. 17394779
Brand: DiscoverX 960044C1
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
The InCELL Hunter™ HEK 293 BLK Kinase Binding Cell Line measures target engagement via compound-induced stabilization of BLK target protein. This cell line ships as two vials of cryopreserved cells.
The InCELL Hunter™ HEK 293 BLK Kinase Binding Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with kinases. This cell line enables assessment of compound-engagement-based stabilization of BLK via detection of BLK levels. This product uses EFC technology. The cell line included overexpresses BLK target protein fused to PL. In the absence of compound that binds and stabilizes the target, this protein is denatured by the heat treatment and no EFC signal is produced. When compound engages and heat-stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Tekniske data
Assay Entry Target Engagement
Cellelinje 293H
Til brug med (applikation) Cell Lines
Mængde 2 hætteglas
Målindgang BLK
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.