Få mere at vide
Tocris Bioscience™ INCB 3284 dimesylate
GREENER_CHOICE
Artikelnummer. 18024412
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18024412 10 mg
18085182 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18024412 Leverandør Tocris Bioscience™ Leverandørnr. 4306/10
Selective CCR2 antagonist
INCB 3284 dimesylate is a potent, selective hCCR2 antagonist. Exhibits potent antagonism against monocyte chemoattractant molecule binding to hCCR2 and chemotaxis activity (IC50 values are 3.7 and 4.7 nM respectively). Also displays weak hERG activity; inhibits the hERG potassium current (IC50 = 84μ M). Orally bioavailable.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 887401-93-6
Molekylær formel C28H39F3N4O10S2
Tekniske data
Kemisk navn eller materiale INCB 3284 dimesylate
Mængde 10 mg
Formel vægt 712.76
Renhed >98%
Molekylvægt (g/mol) 712.753