missing translation for 'onlineSavingsMsg'
Få mere at vide

Inarigivir soproxil, MedChemExpress

Artikelnummer. 16495186
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16495186 100 mg -
16490587 50 mg -
16486977 5 mg -
16474258 10 mg 10mg
16413016 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16495186 Leverandør Medchem Express Leverandørnr. HY109035

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Inarigivir soproxil (SB9200) is an agonist of innate immunity and shows potent antiviral activity against resistant HCV variants, with EC50s of 2.2 and 1.0 μM for HCV 1a/1b in cells of genotype 1 HCV replicon systems. Inarigivir soproxil, an orally bioavailable prodrug of SB 9000, has broad-spectrum antiviral activity against RNA viruses including HCV, norovirus, respiratory syncytial virus and influenza and HBV.

Inarigivir soproxil (SB9200) is an agonist of innate immunity and shows potent antiviral activity against resistant HCV variants, with EC50s of 2.2 and 1.0 μM for HCV 1a/1b in cells of genotype 1 HCV replicon systems. Inarigivir soproxil, an orally bioavailable prodrug of SB 9000, has broad-spectrum antiviral activity against RNA viruses including HCV, norovirus, respiratory syncytial virus and influenza and HBV.

Tekniske data

Kemisk navn eller materiale Inarigivir soproxil
Fysisk form Solid
CAS 942123-43-5
Farve Off-White
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C25H34N7O13PS
Til brug med (applikation) COVID-19-anti-virus
Synonym SB9200 GS-9992
Opløselighedsinformation DMSO : 80 mg/mL (113.70 mM; Need ultrasonic) ∣H2O : 10 mg/mL (14.21 mM; ultrasonic and warming and heat to 60°C)
SMIL CO[C@H]1[C@H](N(C=CC2=O)C(N2)=O)O[C@H](CO)[C@H]1OP(OC[C@H]3O[C@@H](N4C5=NC=NC(N)=C5N=C4)C[C@@H]3O)(SCOC(OC(C)C)=O)=O
Molekylvægt (g/mol) 703.62
Formel vægt 703.62
Procent renhed 98.08%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.