Få mere at vide
STEMCELL Technologies ImmunoCult™ Human CD3/CD28 T Cell Activator
Artikelnummer. 15553062
Encompass
Skift visning
Mængde:
2 ml
5 x 2 ml
Pakningsstørrelse:
2ml
Pakke med 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15553062 2 ml 2ml
16425234 5 x 2 ml Pakke med 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 15553062 Leverandør STEMCELL Technologies Leverandørnr. 10971
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Human T cell activation and expansion reagent
  • Achieve robust activation and expansion of T cells in the absence of magnetic beads, feeder cells, or antigens
  • This product's gentle activation stimulus ensures a high viability of activated T cells, which can be further expanded, for example in ImmunoCult™-XF T Cell Expansion Medium (Catalog #10981)
  • Tetrameric antibody complexes bind to and cross-link CD3 and CD28 cell surface ligands, thereby providing the required signals for T cell activation
  • ImmunoCult™ Human CD3/CD28 T Cell Activator can be used on the Seahorse XF Analyzer to measure T cell activation response and is also available as part of the Agilent Seahorse XF Hu T Cell Activation Assay Kit
  • This product is designed for cell therapy research applications, but may be qualified for use as an ancillary material (AM) following the framework outlined in USP<1043>
  • STEMCELL Technologies can work with you to qualify this reagent as an AM under an approved investigational new drug (IND) or clinical trial application (CTA)
Tekniske data
Celletype T Cells, T Cells, CD4+, T Cells, CD8+
Til brug med (applikation) Activation, Cell Culture, Expansion
Mængde 2 ml