missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Immnoglobulins (IgG, IgA, IgM) Goat anti-Mouse, Affinity Purified, FITC, Secondary Antibody, MP Biomedicals™
Goat Polyclonal Antibody
Brand: MP Biomedicals 0855521
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
- Product is fluorescein-5-isothiocyanate (FITC “Isomer I”)-conjugated goat affinity purified antibody to mouse immunoglobulins (IgG,IgA,IgM) and buffer salts.
- This product is suitable for use as a reagent in immunofluorescence assays (IFA), cell staining (fluorescent microscopy and cell sorting), tissue staining, and blot immunostaining.
- Use of these affinity purified antibodies is recommended to avoid non-specific binding from the inherent antibodies of host animals.
Tekniske data
| Immunohistochemistry, ELISA | |
| FITC | |
| 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, and 10%glycerol (v/v) | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| Liquid |
| Polyclonal | |
| Anti-IgG, IgA, and IgM | |
| Goat | |
| 2 mg | |
| Secondary | |
| Store at 2-8°C. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion