Learn More
Motic™ Immersion Oil, Type HF
Suitable for use with fluorescence.
Brand:  Motic™ 1101001300081
Additional Details : Weight : 0.00900kg
Product Code. 15475252
  • 89.45 DKK / Each
Estimated Shipment: 10-06-2024
to see stock.
Add to basket
Description
Description
  • High refractive index.
  • Transparent.
  • Increases the numerical aperture of the objective lens.
  • Increases resolving power of a microscope.
Specifications
Specifications
Oil immersion objectives/fluorescence
5 mL