missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Imiquimod USP Related Compound A, Mikromol™
Beskrivelse
Imiquimod USP Related Compound A
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Desamino Imiquimod |
| CAS | 99010-24-9 |
| Analyt- eller komponentnavne | Desamino Imiquimod |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H15 N3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1-Isobutyl-1H-imidazo[4,5-c]quinoline (Desaminoimiquimod), 1H-Imidazo[4,5-c]quinoline, 1-(2-methylpropyl)-, 1-(2-Methylpropyl)-1H-imidazo[4,5-c]quinoline, 1-Isobutyl-1H-imidazo-[4,5-c]-quinoline, 1-Isobutyl-1H-imidazo[4,5-c]quinoline, Imiquimod Related Compound A (USP), Imiquimod Related Compound A |
| InChI formel | InChI=1S/C14H15N3/c1-10(2)8-17-9-16-13-7-15-12-6-4-3-5-11(12)14(13)17/h3-7,9-10H,8H2,1-2H3 |
| SMIL | CC(C)Cn1cnc2cnc3ccccc3c12 |
| IUPAC navn | 1-(2-methylpropyl)imidazo[4,5-c]quinoline |
| Vis mere |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?