Få mere at vide
Imipramine N-Oxide, LoGiCal
Artikelnummer. 30459463
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459463 10 mg
1 options
Artikelnummer. 30459463 Leverandør LoGiCal Leverandørnr. LGCFOR0077.12
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Imipramine N-Oxide
CAS 6829-98-7
Analyt- eller komponentnavne Imipramine N-Oxide
Forsendelsestilstand Room Temperature
Molekylær formel C19 H24 N2 O
Certificeringer/compliance ISO 9001
Synonym Imipramine N-Oxide, 5H-Dibenz[b,f]azepine-5-propanamine, 10,11-dihydro-N,N-dimethyl-, N-oxide, 5H-Dibenz[b,f]azepine, 5-[3-(dimethylamino)propyl]-10,11-dihydro-, N-oxide (7CI,8CI), 5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenz[b,f]azepine 5-oxide, Imipraminoxide
InChI formel InChI=1S/C19H24N2O/c1-21(2,22)15-7-14-20-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)20/h3-6,8-11H,7,12-15H2,1-2H3
Løsningstype Neat
SMIL C[N+](C)([O-])CCCN1c2ccccc2CCc3ccccc13
IUPAC navn 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine oxide
Industry Type Clinical Testing
Molekylvægt (g/mol) 296.41
Mængde 10 mg
Formel vægt 296.1889 g/mol