Få mere at vide
Imipramine-d6, TRC
Artikelnummer. 30407432
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30407432 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407432 Leverandør Toronto Research Chemicals Leverandørnr. TRCI4659832.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Imipramine-d6
CAS 65100-45-0
Renhedsgrad noter HPLC
Molekylær formel C19 2H6 H18 N2
Mængde 2.5 mg
Synonym 5H-Dibenz[b,f]azepine-5-propanamine, 10,11-dihydro-N,N-di(methyl-d3)- (9CI), Imipramine-d6, Imipramine-D6 (N,N-dimethyl D6)
InChI formel InChI=1S/C19H24N2/c1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21/h3-6,8-11H,7,12-15H2,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])N(CCCN1c2ccccc2CCc3ccccc13)C([2H])([2H])[2H]
IUPAC navn 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-bis(trideuteriomethyl)propan-1-amine
Molekylvægt (g/mol) 286.44
Formel vægt 286.2316
Procent renhed >95