Få mere at vide
Imipramine-2,4,6,8-d4 Hydrochloride, TRC
Artikelnummer. 30407090
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30407090 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407090 Leverandør Toronto Research Chemicals Leverandørnr. TRCI4659821MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Imipramine-2,4,6,8-d4 Hydrochloride
CAS 61361-33-9
Molekylær formel C19 2H4 H20 N2 . Cl H
Mængde 1 mg
Synonym 5H-Dibenz[b,f]azepine-2,4,6,8-d4-5-propanamine, 10,11-dihydro-N,N-dimethyl-, monohydrochloride (9CI), Imipramine-2,4,6,8-d4 Hydrochloride
InChI formel InChI=1S/C19H24N2.ClH/c1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21;/h3-6,8-11H,7,12-15H2,1-2H3;1H/i3D,4D,10D,11D;
SMIL Cl.[2H]c1cc([2H])c2N(CCCN(C)C)c3c([2H])cc([2H])cc3CCc2c1
IUPAC navn N,N-dimethyl-3-(1,3,8,10-tetradeuterio-5,6-dihydrobenzo[b][1]benzazepin-11-yl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 320.89
Formel vægt 320.1957