Learn More
Imidurea, TRC
Product Code. 30408342
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity Unit Size
30408342 100 g 100g
1 options
Product Code. 30408342 Supplier Toronto Research Chemicals Supplier No. TRCI387570100G
 Restricted Item
Forventet forsendelse: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Imidurea
CAS 39236-46-9
Molecular Formula C11 H16 N8 O8
Quantity 100 g
Synonym Abiol, Biopure 100, Euxyl K 200, Germall 115, Imidazolidinyl urea, Imidurea, Kemipur 100, N,N''-Methylenebis[N'-[3-(hydroxymethyl)-2,5-dioxo-4-imidazolidinyl]urea], Sepicide CI, Urea, N,N''-methylenebis[N'-[3-(hydroxymethyl)-2,5-dioxo-4-imidazolidinyl]-
InChI Formula InChI=1S/C11H16N8O8/c20-2-18-4(6(22)16-10(18)26)14-8(24)12-1-13-9(25)15-5-7(23)17-11(27)19(5)3-21/h4-5,20-21H,1-3H2,(H2,12,14,24)(H2,13,15,25)(H,16,22,26)(H,17,23,27)
SMILES OCN1C(NC(=O)NCNC(=O)NC2N(CO)C(=O)NC2=O)C(=O)NC1=O
IUPAC Name 1-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-3-[[[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoylamino]methyl]urea
Molecular Weight (g/mol) 388.29
Formula Weight 388.1091