missing translation for 'onlineSavingsMsg'
Learn More

Imidocarb Dipropionate, TRC

Product Code. 30404989
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30404989 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404989 Supplier Toronto Research Chemicals Supplier No. TRCI387628100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Imidocarb dipropionate
CAS 55750-06-6
Renhedsgrad noter HPLC
Molekylær formel C19 H20 N6 O . 2 C3 H6 O2
Mængde 100 mg
Synonym Imidocarb Dipropionate, Propanoic acid compd. with N,N'-bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea (2:1), Carbesia, Imidocarb dipropionate, Imizol, Imizol (antiprotozoal), N,N'-Bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea dipropanoate
InChI formel InChI=1S/C19H20N6O.2C3H6O2/c26-19(24-15-5-1-3-13(11-15)17-20-7-8-21-17)25-16-6-2-4-14(12-16)18-22-9-10-23-18;2*1-2-3(4)5/h1-6,11-12H,7-10H2,(H,20,21)(H,22,23)(H2,24,25,26);2*2H2,1H3,(H,4,5)
SMIL CCC(=O)O.CCC(=O)O.O=C(Nc1cccc(c1)C2=NCCN2)Nc3cccc(c3)C4=NCCN4
IUPAC navn 1,3-bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea;propanoic acid
Molekylvægt (g/mol) 496.56
Formel vægt 496.2434
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.