Learn More
Imidazole, Millipore™
Product Code. 10580781
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10580781 250 g 250g
10317190 50 g 50g
10290741 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 10580781 Supplier Millipore Supplier No. 1047160250
 Restricted Item
Forventet forsendelse: 25-11-2026
Buffer substance ACS
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 288-32-4
Specifications
Melting Point 90°C
Boiling Point 256°C/1013 hPa
Density 1.233 g/cm3 at 20°C
Quantity 250 g
MDL Number MFCD00005183
Synonym Imidazole, Glyoxaline, Iminazole, Imidazole
Molecular Weight (g/mol) 68.08
Grade Research
Chemical Name or Material Imidazole