Learn More
Thermo Scientific Chemicals Imidazole, 0.5M buffer soln., pH 7.0
Catalog Number p-7042458
Click to view available options
Quantity:
100 mL
250 mL
288-32-4
288-32-4
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Concentration 0.5M buffer soln.
Molecular Weight (g/mol) 68.079, 68.08
pH 7.0
Molecular Formula C3H4N2
Quantity 100 mL
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
SMILES N1C=CN=C1
IUPAC Name 1H-imidazole
Content And Storage Keep cold
Form Liquid