Få mere at vide
Imidazo[5,1-b]thiazole-3-carboxylic Acid Hydrochloride, TRC
Artikelnummer. 30408505
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30408505 10 mg
30404032 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30408505 Leverandør Toronto Research Chemicals Leverandørnr. TRCI70785010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2137605-31-1
Molekylær formel C6H4N2O2S . HCl
Molekylvægt (g/mol) 204.63
Synonym Imidazo[4,3-b][1,3]thiazole-3-carboxylic acid hydrochloride
IUPAC navn imidazo[5,1-b][1,3]thiazole-3-carboxylic acid;hydrochloride
SMIL Cl.OC(=O)c1csc2cncn12
Tekniske data
Kemisk navn eller materiale Imidazo[5,1-b]thiazole-3-carboxylic Acid Hydrochloride
Mængde 10 mg
InChI formel InChI=1S/C6H4N2O2S.ClH/c9-6(10)4-2-11-5-1-7-3-8(4)5;/h1-3H,(H,9,10);1H
Formel vægt 203.976