Få mere at vide
Imidaprilat-d3, TRC
Artikelnummer. 30138049
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138049 1 mg
17989339 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30138049 Leverandør Toronto Research Chemicals Leverandørnr. TRCI2750521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1356019-69-6
Molekylær formel C18H20D3N3O6
Molekylvægt (g/mol) 380.41
Synonym (4S)-3-{(2S)-2-[N-((1S)-1-Carboxy-3-phenyl-d3-propyl)amino]propionyl}-1-methyl-2-oxoimidazolidine-4-carboxylic Acid, Imidapril Diacid-d5;
SMIL C[C@@H](C(N1[C@@H](C(O)=O)CN(C([2H])([2H])[2H])C1=O)=O)N[C@H](C(O)=O)CCC2=CC=CC=C2
Tekniske data
Kemisk navn eller materiale Imidaprilat-d3
Mængde 1 mg
InChI formel InChI=1S/C18H23N3O6/c1-11(15(22)21-14(17(25)26)10-20(2)18(21)27)19-13(16(23)24)9-8-12-6-4-3-5-7-12/h3-7,11,13-14,19H,8-10H2,1-2H3,(H,23,24)(H,25,26)/t11-,13-,14+/m0/s1/i2D3