Learn More
IMD-0354, MedChemExpress
Product Code. 16463528
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16463528 10 mg 10mg
16406257 5 mg 5mg
16439966 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16463528 Supplier Medchem Express Supplier No. HY10172
 Restricted Item
Forventet forsendelse: 30-11-2026
MedChemExpress IMD-0354 (IKK2 Inhibitor V) is a selective IKKβ inhibitor which inhibits NF-κB activity. IMD0354 inhibits TNF-α induced NF-κB transcription activity with an IC50 of 1.2 uM.
IMD-0354 (IKK2 Inhibitor V) is a selective IKKβ inhibitor which inhibits NF-κB activity. IMD0354 inhibits TNF-α induced NF-κB transcription activity with an IC50 of 1.2 uM.
Tekniske data
Chemical Name or Material IMD-0354
CAS 978-62-1
Color White
Purity Grade Notes Research
Molecular Formula C15H8ClF6NO2
Quantity 10 mg
Synonym IKK2 Inhibitor V
Solubility Information DMSO : ≥ 100 mg/mL (260.64 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(NC1=CC(C(F)(F)F)=CC(C(F)(F)F)=C1)C2=CC(Cl)=CC=C2O
Molecular Weight (g/mol) 383.67
Formula Weight 383.67
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease