Få mere at vide
Imazalil-d5 Sulfate, TRC
Artikelnummer. 30403066
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30403066 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403066 Leverandør Toronto Research Chemicals Leverandørnr. TRCI26850225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Imazalil-d5 Sulfate
CAS 1398065-92-3
Molekylær formel C14 D5 H9 Cl2 N2 O . H2 O4 S
Mængde 25 mg
Synonym 1-[2-(2,4-Dichlorophenyl)-2-(1,1,2,3,3-pentadeuterioprop-2-enoxy)ethyl]imidazole sulfuric acid, Imazalil-d5 Sulfate, Imazalil-D5 (2-propenyl-D5) Sulfate, (±)-Imazalil-d5 Sulfate (allyloxy-d5), Enilconazole-D5 (allyloxy-D5) Sulfate, Enilconazole-d5 Sulfate, Chloramizol-d5 sulfate, 1-[2-(2,4-Dichlorophenyl)-2-(2-propenyloxy-1,1,2,3,3-d5)ethyl]-1H-imidazole sulfate (1:1)
InChI formel InChI=1S/C14H14Cl2N2O.H2O4S/c1-2-7-19-14(9-18-6-5-17-10-18)12-4-3-11(15)8-13(12)16;1-5(2,3)4/h2-6,8,10,14H,1,7,9H2;(H2,1,2,3,4)/i1D2,2D,7D2;
SMIL [2H]C(=C([2H])C([2H])([2H])OC(Cn1ccnc1)c2ccc(Cl)cc2Cl)[2H].OS(=O)(=O)O
IUPAC navn 1-[2-(2,4-dichlorophenyl)-2-(1,1,2,3,3-pentadeuterioprop-2-enoxy)ethyl]imidazole;sulfuric acid
Molekylvægt (g/mol) 400.289
Formel vægt 399.047