Få mere at vide
Tocris Bioscience™ Imatinib mesylate
Potent and selective v-Abl tyrosine kinase inhibitor; also inhibits PDGFR and c-kit
Brand: Tocris Bioscience™ 5906/100
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Imatinib mesylate is a potent and selective v-Abl tyrosine kinase inhibitor (IC50 = 38 nM). Also inhibits PDGFR and c-kit. Exhibits selectivity for v-Abl over a panel of other tyrosine and serine/threonine protein kinases. Selectively inhibits PDGF-stimulated growth of v-abl-transformed PB-3 cells and v-sis-transformed BALB/c 3T3 cells in vitro. Exhibits antitumor effects in mice bearing AMuLV or BABL/c 3T3 v-sis cells. Inhibits replication of SARS-CoV and MERS-CoV in vitro (EC50 values are 9.823 and 17.689 μM, respectively).Tekniske data
| 4-[(4-Methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide methanesulfonate | |
| 220127-57-1 | |
| Gleevec, CGP 57148B | |
| 100 mg |