Få mere at vide
Imatinib 4-Benzoic Acid, TRC
Artikelnummer. 30479979
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479979 10 mg
30473517 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30479979 Leverandør Toronto Research Chemicals Leverandørnr. TRCM24676010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Imatinib 4-Benzoic Acid
CAS 1356565-49-5
Molekylær formel C36 H35 N7 O3
Mængde 10 mg
Synonym 4-[[4-[[4-[[[4-Methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]amino]carbonyl]phenyl]methyl]-1-piperazinyl]methyl]benzoic Acid, 4-((4-(4-((4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)carbamoyl)benzyl)piperazin-1-yl)methyl)benzoic Acid
InChI formel InChI=1S/C36H35N7O3/c1-25-4-13-31(21-33(25)41-36-38-16-14-32(40-36)30-3-2-15-37-22-30)39-34(44)28-9-5-26(6-10-28)23-42-17-19-43(20-18-42)24-27-7-11-29(12-8-27)35(45)46/h2-16,21-22H,17-20,23-24H2,1H3,(H,39,44)(H,45,46)(H,38,40,41)
SMIL Cc1ccc(NC(=O)c2ccc(CN3CCN(Cc4ccc(cc4)C(=O)O)CC3)cc2)cc1Nc5nccc(n5)c6cccnc6
IUPAC navn 4-[[4-[[4-[[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]carbamoyl]phenyl]methyl]piperazin-1-yl]methyl]benzoic acid
Molekylvægt (g/mol) 613.708
Formel vægt 613.28