missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Image-iT™ Low Density Lipoprotein Uptake Kit, pHrodo™ Red

The Image-iT Low Density Lipoprotein Uptake Kit - pHrodo Red includes pHrodo Red dye-labeled LDL (also available separately), unlabeled LDL, heparin, BSA, and assay buffer.

Brand:  Invitrogen™ I34360

3220.00 DKK valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 15739230

  • 4730.00 DKK / 1 set

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The kit was designed to specifically detect binding and uptake of LDL through the LDL receptor internalization pathway, allowing maximum control and flexibility in the experimental design. It can be used with fluorescence microscopy, flow cytometry, cell sorting, high-throughput analysis, and high content analysis.

• pHrodo Red-LDL is detectable with RFP/TRITC filter set (absorption peak: ∼560 nm, emission peak: ∼585 nm)
• pHrodo Red-LDL is multiplexable with green, deep red, violet, and blue emission dyes, such as GFP, DAPI, Hoechst, HCS LipidTOX, and LysoTracker Deep Red
• No-wash protocol is possible, allowing dynamic measurement of LDL uptake and trafficking

pHrodo Red- LDL is dimly fluorescent at neutral pH outside of cells, but fluoresces brightly after endocytosis. This feature allows for a no-wash protocol when using pHrodo Red-LDL, enabling kinetic measurement of LDL uptake, endosomal maturation, and trafficking in cells.

Two controls are included in the kit: unlabeled LDL as an optional pretreatment to occlude cell surface receptors in advance of probing with the labeled construct, and heparin which will chelate the LDL in the solution phase outside of the cells, preventing binding and uptake. When cells are labeled in the absence of these pretreatment controls, a fluorescent signal indicates specific LDL receptor-mediated endocytosis.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

  • 1 x 200 μL pHrodo Red-LDL, 1 mg/mL
  • 1 x 200 μL LDL (unlabeled), 2.5 mg/mL
  • 1 x 50 mL LDL assay buffer, 1 X
  • 1 vial heparin, 5 mg/mL
  • 1 x 150 mg BSA
  • Store in refrigerator (2°C to 8°C) and protect from light.
pH-Sensitive Dyes
1 kit
Lipoprotein Uptake Kit
Molecular Probes
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3011682 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog NumberI34360
Lot Number2901488 Certificate TypeCertificate of Analysis Date03/04/2024 Catalog NumberI34360
Lot Number2738244 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog NumberI34360
Lot Number2599178 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberI34360
Lot Number2552924 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog NumberI34360
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Image-iT™ Low Density Lipoprotein Uptake Kit, pHrodo™ Red > 1 x 200 μL pHrodo Red-LDL, 1 mg/mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.