missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT6/CD85e/LILRA3 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-00150AF405
Additional Details : Weight : 0.00970kg
Description
ILT6/CD85e/LILRA3 Polyclonal specifically detects ILT6/CD85e/LILRA3 in Human samples. It is validated for ELISA.Specifications
ILT6/CD85e/LILRA3 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
11026 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 405 | |
CD85 antigen-like family member E, CD85e antigen, HM31, ILT-6, ILT6CD85e, Immunoglobulin-like transcript 6, leucocyte immunoglobulin-like receptor, Leukocyte immunoglobulin-like receptor 4, leukocyte immunoglobulin-like receptor A3, leukocyte immunoglobulin-like receptor subfamily A member 3, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member3, LIR4CD85E, LIR-4HM43e3, Monocyte inhibitory receptor HM43/HM31 | |
Produced in rabbits immunized with purified, recombinant ILT6/CD85e/LILRA3 (AAH28208.1; Met1-Glu439). | |
0.1 mL | |
Primary | |
Human | |
Purified |