missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT3/CD85k/LILRB4 Antibody (Merck patent anti-ILT3 complex), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28105CL1
This item is not returnable.
View return policy
Description
ILT3/CD85k/LILRB4 Monoclonal antibody specifically detects ILT3/CD85k/LILRB4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
ILT3/CD85k/LILRB4 | |
Monoclonal | |
CoraFluor 1 | |
CD85 antigen-like family member K, CD85k, CD85k antigen, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5LILRB5, LIR-5subfamily B, member 4, member 4 | |
LILRB4/ ILT3/ CD85k | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Functional Assay | |
Merck patent anti-ILT3 complex | |
PBS | |
Human | |
Protein A purified | |
RUO | |
11006 | |
Store at 4°C in the dark. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ILT3/CD85k/LILRB4 Antibody (Merck patent anti-ILT3 complex), CoraFluor™ 1, Novus Biologicals™ > 0.1 mL; CoraFluor 1
Spot an opportunity for improvement?Share a Content Correction