missing translation for 'onlineSavingsMsg'
Learn More

IL8 Recombinant Human Protein, hIgG1-Fc Tag

IL8 Recombinant Human Protein, hIgG1-Fc Tag

Brand:  Invitrogen 10098H01H2U5

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15294635

  • 431.00 DKK / 5µg


Estimated Shipment
13-08-2020
Add to basket

Description

Description

Interleukin 8 (IL8), recombinant human protein is supplied as a lyophilized powder.

This recombinant protein was expressed from a DNA sequence encoding the 72 amino acid residue form (Ser 28-Ser 99) of mature human IL8 (NP_000575.1) fused to the Fc region of human IgG1 at the N-terminus.

Activity: This recombinant protein has not been tested.

Formulation: 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

Specifications

Specifications

Mammalian
Assay standard
Lyophilized
3576
IL8 Recombinant Human Protein
≥85% by SDS-PAGE
–20°C
1 Each
IL8 Recombinant Human Protein
IL8
Human
Chemokines and receptors
Protein assays and analysis
Lyophilized
35kDa
PAGE
5μg
For research use only. Not for use in diagnostic procedures
CXCL8, GCP-1, GCP1, LECT, LUCT, LYNAP, MDNCF, MONAP, NAF, NAP-1, NAP1
1 Vial of lyophilized protein
IL8 Recombinant Human Protein
IgG1 Fc Tag, hIgG1-Fc Tag
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.