missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL6R Antibody (rhPM-1 (Tocilizumab)), Alexa Fluor™ 594, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP2-75192AF594
Additional Details : Weight : 0.00970kg
Description
IL6R Monoclonal specifically detects IL6R in Human samples. It is validated for ELISA, Immunofluorescence.Specifications
IL6R | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Human | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunofluorescence | |
rhPM-1 (Tocilizumab) | |
ELISA, Immunofluorescence | |
CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
This antibody was prepared by CDR grafting of mouse AUK 12-20 antibody onto a human framework regions. | |
0.1 mL | |
Cancer, Cytokine Research, Immunology, Innate Immunity | |
3570 | |
Store at 4°C in the dark. | |
IgG1 κ |