Learn More
Invitrogen™ IL6 (Interleukin 6); Recombinant Human Protein
Recombinant Human Interleukin 6 (IL6)
10210.00 DKK - 49620.00 DKK
Specifications
Molecular Weight (g/mol) | 21.3 kDa |
---|---|
Purification Method | Sequential Chromatography |
Gene Alias | IL-6 (Interleukin 6) |
Protein Length | 184 aa |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11370832
|
Invitrogen™
CTP0061 |
100 μg |
10210.00 DKK
Each |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11380832
|
Invitrogen™
CTP0063 |
1 mg |
49620.00 DKK
Each |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The lyophilized, E. coli-expressed IL6 CTS™ Recombinant Human is verified for activity through dose-dependent proliferation of mouse B9 cells.
- Qualified—helps reduce your burden in qualifying reagents for regulatory submission in cell therapy applications
- Highly Pure—minimizes interference from other proteins or contaminants
- Active—get more results with less protein
Simplified Transition into Clinical Research
GIBCO™ Cell Therapy Systems (CTS™) products are of high quality and supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate. The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.
High Purity
To help verify that GIBCO™ CTS™ Recombinant growth factors are of the highest quality, each protein is analyzed for purity, along with structural homogeneity, to help ensure a biologically active protein. The purity of each GIBCO™ CTS™ recombinant growth factor is verified by SDS-PAGE to be over 95% pure and to have endotoxin levels below 0.1 ng/μg.
High Biological Activity
All GIBCO™ CTS™ Recombinant growth factors are verified for activity. Each lot has its own ED50 determined using an activity assay based on a specific function of the growth factor. The activity of our IL6 CTS™ Recombinant Human growth factor is determined by the dose-dependent proliferation of mouse B9 cells to have an ED50 range of 0.008–0.02ng/mL (specific activity: 1.25 × 108–5.0 × 107 units/mg). Optimal concentration for individual applications should be determined by a dose response assay.
Specifications
21.3 kDa | |
IL-6 (Interleukin 6) | |
Human | |
Cytokines & Receptors | |
95 % |
Sequential Chromatography | |
184 aa | |
Recombinant | |
Interleukins |
For Research Use Only. Not for use in diagnostic procedures.