missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IL11 (Human) Recombinant Protein

Human IL11 full-length ORF ( NP_000632, 1 a.a. - 199 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00003589-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180075

  • 2735.00 DKK / 10µg
Estimated Shipment: 19-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MNCVCRLVLVVLSLWPDTAVAPGPPPGPPRVSPDPRAELDSTVLLTRSLLADTRQLAAQLRDKFPADGDHNLDSLPTLAMSAGALGALQLPGVLTRLRADLLSYLRHVQWLRRAGGSSLKTLEPELGTLQARLDRLLRRLQLLMSRLALPQPPPDPPAPPLAPPSSAWGGIRAALAILGGLHLTLDWAVRGLLLLKTRL
Specifications

Specifications

NP_000632
interleukin 11
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IL11
GST
3589
Wheat germ expression system
10 μg
AGIF, IL-11
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers