missing translation for 'onlineSavingsMsg'
Learn More

IL-9 Receptor Alpha (CD129) Mouse anti-Human, BV650, Clone: AH9R7, BD OptiBuild™

Mouse Monoclonal Antibody

Brand:  BD Biosciences 745308

3684.01 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15809699

  • 3685.00 DKK / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IL-9 Receptor Alpha (CD129) Monoclonal antibody specifically detects IL-9 Receptor Alpha (CD129) in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-9 Receptor Alpha (CD129)
Monoclonal
0.2 mg/mL
Mouse
RUO
Human
Purified
Flow Cytometry
AH9R7
Brilliant Violet 650
50 ÎĽg
Primary
Refrigerate (2 to 8°C)
IgG2b Îş
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IL-9 Receptor Alpha (CD129) Mouse anti-Human, BV650, Clone: AH9R7, BD OptiBuild™ > Hu IL-9R (CD129) BV650 AH9R7 50 μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.