Få mere at vide
Novus Biologicals™ IL-9 Overexpression Lysate
Artikelnummer. 18267184 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mg
Artikelnummer. Mængde
18267184 0.1 mg
1 options
Artikelnummer. 18267184 Leverandør Novus Biologicals™ Leverandørnr. NBL111970
 Begrænset vare
Forventet forsendelse: 15-10-2026
The IL-9 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Tekniske data
Cellelinje The lysate was created in HEK293T cells, using plasmid ID RC29682 and based on accession number NM_59. The protein contains a C-terminal DDK tag.
Til brug med (applikation) Western Blot
Mængde 0.1 mg
Opbevaringsbuffer RIPA buffer
Renhed Protein
Testspecificitet Homo sapiens interleukin 9 (IL9), mRNA.
Indhold og opbevaring -80°C. Avoid freeze-thaw cycles.
Proteintilstand Native
For Research Use Only