missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
IL-7R alpha/CD127 Antibody (YIL7R323), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81081AF405
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Product Image.
Tekniske data
| IL-7R alpha/CD127 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| IL7R | |
| Recombinant monoclonal antibody to IL-7R alpha/CD127r. | |
| 0.1 mL | |
| Apoptosis, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
| 3575 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| YIL7R323 | |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody binds to an epitope of human IL-7R alpha/CD127 distinct to that bound by Ab00531. | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction