Learn More
IL-6R alpha Antibody (5), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90511H
Additional Details : Weight : 0.00970kg
Description
IL-6R alpha Monoclonal specifically detects IL-6R alpha in Mouse samples. It is validated for ELISA.Specifications
IL-6R alpha | |
Monoclonal | |
HRP | |
CD126, gp80, IL6Q, IL6R, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-6R alpha (NP_034689.2; Ile 44-Glu 357). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Human IL6R/ CD126 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
5 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
3570 | |
Mouse | |
Purified |