missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
IL-22 Antibody (fezakinumab), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28333CL1
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IL-22 Monoclonal antibody specifically detects IL-22 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
| IL-22 | |
| Monoclonal | |
| CoraFluor 1 | |
| Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
| IL-22 | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research, Innate Immunity | |
| 50616 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| fezakinumab | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion