missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
IL-17F Antibody (8F5.1A9), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-12034AF405
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IL-17F Monoclonal antibody specifically detects IL-17F in Mouse samples. It is validated for Flow Cytometry
Tekniske data
| IL-17F | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| IgG κ |
| Flow Cytometry | |
| 8F5.1A9 | |
| Flow Cytometry | |
| Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F | |
| This antibody was raised by immunising mice with murine IL-17F coupled to ovalbumin to generate a highly immunogenic complex. | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 112744 | |
| Store at 4°C in the dark. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion