Learn More
Invitrogen™ IL-11 Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMIL11X5
Additional Details : Weight : 0.33800kg
Description
The Mouse Interleukin-11(Ms IL-11) ELISA quantitates Ms IL-11 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IL-11. Principle of the method The Mouse IL-11 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Interleukin 11 is a pleiotropic cytokine produced by mesenchymal-derived adherent cells. IL11 shares many functions of IL6 and LIF, including potentiation of megakaryocyte activity, enhancement of human myeloma cell proliferation, and enhancement of hepatic acute phase protein production.Interleukin 11 directly stimulates the proliferation of hematopoietic stem cells and megakaryocyte progenitor cells, and induces megakaryocyte maturation resulting in increased platelet production. Clinically it is used to prevent severe thrombocytopenia and the reduction of the need for platelet transfusion following myelosuppressive chemotherapy.Specifications
P47873 | |
1000 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
16156 | |
<12% | |
Chromogen, SAV-HRP, Pre-coated 96 well plates, Stop Solution, Biotinylated Detection Antibody, Wash Buffer, Adhesive Plate Covers, Assay Diluent Concentrate, Standard | |
5 x 96 Tests | |
Cell Cycle & Proliferation | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
1.45 ng/mL to 450 ng/mL | |
1 ng/mL to 450 ng/mL | |
Biotin | |
Other Proteins | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-13, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-5, M-CSF, MIG, MIP- 1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF RI, TPO, VEGF. | |
Mouse | |
Colorimetric Microplate Reader | |
IL-11 | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
IL-11 | |
1 hr. 20 min. |